Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DiGeorge syndrome critical region gene 6, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human DGCR6 protein.

Manufacturer:  Abnova Corporation H00008214B01

Catalog No. 89-006-548


Add to cart

Description

Description

DiGeorge syndrome, and more widely, the CATCH 22 syndrome, are associated with microdeletions in chromosomal region 22q11.2. The product of this gene shares homology with the Drosophila melanogaster gonadal protein, which participates in gonadal and germ cell development, and with the gamma-1 subunit of human laminin. This gene is a candidate for involvement in DiGeorge syndrome pathology and in schizophrenia. [provided by RefSeq

Sequence: MERYAGALEEVADGARQQERHYQLLSALQSLVKELPSSFQQRLSYTTLSDLALALLDGTVFEIVQGLLEIQHLTEKSLYNQRLRLQNEHRVLRQALRQKHQEAQQACRPHNLPVLQAAQQRELEAVEHRIREEQRAMDQKIVLELDRKVADQQSTLEKAGVAGFYVTTNPQELMLQMNLLELIRKLQQRGCWAGKAALGLGGPWQLPAAQCDQKGSPVPP
Specifications

Specifications

DiGeorge syndrome critical region gene 6
Unconjugated
No additive
NM_005675.3
DGCR6
DGCR6 (NP_005666.2, 1 a.a. ~ 220 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
8214
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human DGCR6 protein.
DGCR6
Serum
Mouse
50 uL
Cell Adhesion
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit