Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGCR6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526613
Description
DGCR6 Polyclonal antibody specifically detects DGCR6 in Human, Mouse samples. It is validated for Western Blot.Specifications
DGCR6 | |
Polyclonal | |
Unconjugated | |
DGCR6 | |
Dgcr6; DiGeorge syndrome chromosome region 6; DiGeorge syndrome critical region 6; DiGeorge syndrome critical region 6 homolog; DiGeorge syndrome critical region gene 6; DiGeorge syndrome critical region protein 6; protein DGCR6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
13353, 8214 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O35347, Q14129 | |
DGCR6 | |
KLH conjugated synthetic peptide between 103-129 amino acids from the central region of human DGCR6 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction