Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGKθ Mouse, Unlabeled, Clone: 24, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610930

 View more versions of this product

Catalog No. BDB610930


Add to cart

Description

Description

The protein kinase C pathway is a major signal transduction system that is activated upon stimulation of transmembrane receptors by hormones, neurotransmitters, and growth factors. Key mediators in this pathway are increased intracellular free Ca2+ levels and formation of diacylglycerol (DAG). DGKθ (diacylglycerol kinase θ) restricts PKC activation through the phosphorylation of DAG molecules that contain an unsaturated fatty acid at the sn-2 position to produce phosphatidic acid (PA). DGKθ contains several regions that are found in signaling molecules where they function in lipid-protein and protein-protein interactions. A C-terminal catalytic domain, three CRDs (cysteine rich domains), a PH domain, and an N-terminal proline/glycine rich domain are structural features of DGKθ. Six potential PKC phosphorylation sites lie between CRD3 and the PH domain. Cell-specific expression differentiate multiple isoforms of DGK. DGKθ is expressed primarily within the cerebellar cortex and hippocampus of the brain, but is also found in the small intestine and liver. The presence of the RA (Ras-associating) domain suggests that DGKθ may mediate activity of the Ras-like small GTP binding proteins.
Specifications

Specifications

DGKθ
24
Unconjugated
Affinity Purified
Human DGKθ aa. 677-883
50μg
Cell Biology
Primary
Human, Murine, Rabbit, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.