Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGKA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515418

Catalog No. PIPA515418


Add to cart

Description

Description

Upon cell stimulation, the kinase DGKA converts the second messenger diacylglycerol into phophatidate, initiating the resynthesis of phosphatidylinositols and attenuating protein kinase C activity. DGKA is stimulated by calcium and phosphatidylserine, and is phosphorylated by protein kinase C. Tissue expression is in lymphocytes and oligodengroglial cells. DGKA contains 2 zinc-dependent phorbol-ester and DAG binding domains, and 2 EF-hand calcium binding domains.
Specifications

Specifications

DGKA
Polyclonal
Unconjugated
DGKA
Liquid
DGKA
KLH conjugated synthetic peptide between 598-628 amino acids from the C-terminal region of human DGKA
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide
P23743
Diacylglycerol kinase alpha; Diglyceride kinase; DGK-alpha; DAG kinase alpha; 80 kDa diacylglycerol kinase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1606
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit