Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

diacylglycerol kinase, gamma 90kDa, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human DGKG protein.

Manufacturer:  Abnova Corporation H00001608B01

Catalog No. 89-005-223


Add to cart

Description

Description

This gene encodes an enzyme that is a member of the type I subfamily of diacylglycerol kinases, which are involved in lipid metabolism. These enzymes generate phosphatidic acid by catalyzing the phosphorylation of diacylglycerol, a fundamental lipid second messenger that activates numerous proteins, including protein kinase C isoforms, Ras guanyl nucleotide-releasing proteins and some transient receptor potential channels. Diacylglycerol kinase gamma has been implicated in cell cycle regulation and in the negative regulation of macrophage differentiation in leukemia cells. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MGEERWVSLTPEEFDQLQKYSEYSSKKIKDALTEFNEGGSLKQYDPHEPISYDVFKLFMRAYLEVDLPQPLSTHLFLAFSQKPRHETSDHPTEGASNSEANSADTNIQNADNATKADEACAPDTESNMAEKQAPAEDQVAATPLEPPVPRSSSSESPVVYLKDVVCYLSLLETGRPQDKLEFMFRLYDSDENGLLDQAEMDCIVNQMLHIAQYLEWDPTELRPILKEMLQGMDYDRDGFVSLQEWVHGGMTTIPLLVLLGMDDSGSKGDGRHAWTMKHFKKPTYCNFCHIMLMGVRKQGLCCTYCKYTVHERCVSRNIPGCVKTYSKAKRSGEVMQHAWVEGNSSVKCDRCHKSIKCYQSVTARHCVWCRMTFHRKCELSTLCDGGELRDHILLPTSICPITRDRPGEKSDGCVSAKGELVMQYKIIPTPGTHPLLVLVNPKSGGRQGERLNFFRDTPDFRVLACGGDGTVGWILDCIDKANFAKHPPVAVLPLGTGNDLARCLRWGGGYEGGSLTKILKDIEQSPLVMLDRWHLEVIPREEVENGDQVPYSIMNNYFSIGVDASIAHRFHVMREKHPEKFNSRMKNKLWYFEFGTSETFAATCKKLHDHIELECDGVGVDLSNIFLEGIAILNIPSMYGGTNLWGENKKNRAVIRESRKGVTDPKELKFCVQDLSDQLLEVVGLEGAMEMGQIYTGLKSAGRRLAQCASVTIRTNKLLPMQVDGEPWMQPCCTIKITHKNQAPMMMGPPQKSSFFSLRRKSRSKD
Specifications

Specifications

diacylglycerol kinase, gamma 90kDa
Unconjugated
No additive
BC089411
DAGK3|DGK-GAMMA|MGC104993|MGC133330
Mouse
50 uL
Endocrine/Metabolism
Y
Polyclonal
Human, Rat
Western Blot
Mouse polyclonal antibody raised against a full-length human DGKG protein.
DGKG
Serum
DGKG
DGKG (NP_001074213.1, 1 a.a. ~ 766 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1608
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit