Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGKK, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA525046
Description
DGKK phosphorylates diacylglycerol (DAG) to generate phosphatidic acid (PA).
Specifications
DGKK | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q5KSL6 | |
142kDa diacylglycerol kinase, DAG kinase kappa, DGK-kappa, diacylglycerol kinase kappa, diglyceride kinase kappa | |
Rabbit | |
IgG | |
400μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
139189, 331374 |
Immunohistochemistry, Western Blot | |
Unconjugated | |
DGKK | |
Liquid | |
DGKK | |
KLH conjugated synthetic peptide between 1232-1261 amino acids from the C-terminal region of human DGKK | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Murine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title