Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHFR Mouse, Unlabeled, Clone: 49, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610696

 View more versions of this product

Catalog No. BDB610696


Add to cart

Description

Description

Dihydrofolate reductase (DHFR) regenerates tetrahydrofolate from dihydrofolate in the presence of NADPH. Tetrahydrofolate is the coenzyme for thymidilate synthetase in the biosynthesis of thymidine and is also critical for the synthesis of amino acids and purines. DHFR is a protein of 186 amino acids that is highly conserved among different organisms. DHFR levels change during the cell cycle, with the highest content during the G1/S transition. Expression of DHFR tightly correlates with the turnover rate of its mRNA. Because dividing cells require a continuous supply of thymidine, DHFR has been a target for anticancer drugs. The folic acid antagonists aminopterin and amethopterin (methotrexate), widely used in cancer treatments, inhibit DHFR. However, over long treatments, treated cells often amplify the DHFR gene, producing drug-resistant cells.

Specifications

Specifications

DHFR
49
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.