Learn More
DHPS, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DHPS.
Manufacturer: Abnova Corporation PAB25205
Description
The unusual amino acid hypusine is formed posttranslationally and is only found in a single cellular protein, eukaryotic translation initiation factor 5A. In the first step of hypusine biosynthesis, deoxyhypusine synthase catalyzes the NAD-dependent transfer of the butylamine moiety of spermidine to the epsilon-amino group of a specific lysine residue of the EIF5A precursor protein to form the intermediate deoxyhypusine residue. This gene consists of nine exons spanning 6.6kb. Three transcript variants have been isolated. However, only transcript variant 1 encodes an active protein. The shorter variants may act as modulating factors of DHPS activity. [provided by RefSeq
Specifications
DHPS | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of DHPS. | |
Liquid | |
DHPS | |
1725 | |
Rabbit | |
Polyclonal | |
Affinity purification | |
RUO | |
Human |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
MIG13 | |
DHPS | |
A synthetic peptide corresponding to DHPS. | |
Primary | |
100 uL | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
DHPS polyclonal antibody detects endogenous levels of DHPS protein. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only