Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

deoxyhypusine synthase, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human DHPS protein.

Manufacturer:  Abnova Corporation H00001725B02

Catalog No. 89-005-243


Add to cart

Description

Description

The unusual amino acid hypusine is formed posttranslationally and is only found in a single cellular protein, eukaryotic translation initiation factor 5A. In the first step of hypusine biosynthesis, deoxyhypusine synthase catalyzes the NAD-dependent transfer of the butylamine moiety of spermidine to the epsilon-amino group of a specific lysine residue of the EIF5A precursor protein to form the intermediate deoxyhypusine residue. This gene consists of nine exons spanning 6.6kb. Three transcript variants have been isolated. However, only transcript variant 1 encodes an active protein. The shorter variants may act as modulating factors of DHPS activity. [provided by RefSeq

Sequence: MEGSLEREAPAGALAAVLKHSSTLPPESTQVRGYDFNRGVNYRALLEAFGTTGFQATNFGRAVQQVNAMIEKKLEPLSQDEDQHADLTQSRRPLTSCTIFLGYTSNLISSGIRETIRYLVQHNMVDVLVTTAGGVEEDLIKCLAPTYLGEFSLRGKELRENGINRIGNLLVPNENYCKFEDWLMPILDQMVMEQNTEGVKWTPSKMIARLGKEINNPESVYYWAQKNHIPVFSPALTDGSLGDMIFFHSYKNPGLVLDIVEDLRLINTQAIFAKCTGMIILGGGVVKHHIANANLMRNGADYAVYINTAQEFDGSDSGARPDEAVSWGKIRVDAQPVKVYADASLVFPLLVAETFAQKMDAFMHEKNED
Specifications

Specifications

deoxyhypusine synthase
Unconjugated
No additive
NM_001930
MIG13
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human DHPS protein.
DHPS
Serum
DHPS
DHPS (NP_001921, 1 a.a. ~ 369 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1725
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit