Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DHPS, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232256

Catalog No. NBP232256


Add to cart

Description

Description

DHPS Polyclonal antibody specifically detects DHPS in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

DHPS
1.0 mg/mL
Western Blot 1:500 - 1:2500, Immunoprecipitation 2 - 10 ug/mg lysate
NP_001921.1
deoxyhypusine synthase, DHS, DS, EC 2.5.1, EC 2.5.1.46, MIG13, migration-inducing gene 13
Rabbit
IgG
100 ul
Cell Cycle and Replication
Primary
1725
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
DHPS
The immunogen this antibody was made to, maps to a region between residue 319 to 369 of human Deoxyhypusine Synthase using the numbering given in entry NP_001921.1 (GeneID 1725).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only