Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHRS7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524706

Catalog No. PIPA524706

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DHRS7 is short-chain dehydrogenases/reductases (SDRs), such as DHRS7, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids.
Specifications

Specifications

DHRS7
Polyclonal
PBS with 0.09% sodium azide
Q9Y394
SDR34C1, retDSR4, retSDR4, dehydrogenase/reductase SDR family member 7, retinal short-chain dehydrogenase/reductase 4, short chain dehydrogenase/reductase family 34C, member 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51635
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DHRS7
Liquid
DHRS7
KLH conjugated synthetic peptide between 66-93 amino acids from the N-terminal region of human DHRS7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit