Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DHX30, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126603

Catalog No. NBP126603


Add to cart

Description

Description

DHX30 Polyclonal antibody specifically detects DHX30 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

DHX30
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 5-10 ug/mg lysate
NP_619520.1
ATP-dependent RNA helicase DHX30, DDX30, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 30, DEAH (Asp-Glu-Ala-His) box polypeptide 30, DEAH box protein 30, EC 3.6.1, EC 3.6.4.13, FLJ11214, KIAA0890RETCOR, putative ATP-dependent RNA helicase DHX30, retina co-repressor
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
DHX30
The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human DEAH (Asp-Glu-Ala-His) box polypeptide 30 using the numbering given in entry NP_619520.1 (GeneID 22907).
Immunogen affinity purified
RUO
Primary
22907
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only