Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIS3L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525525

Catalog No. PIPA525525


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

DIS3L is a probable exonuclease. It has four isoforms.
Specifications

Specifications

DIS3L
Polyclonal
PBS with 0.09% sodium azide
Q8TF46, Q8C0S1
DIS3L1, DIS3-like exonuclease 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
115752, 213550
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DIS3L
Liquid
DIS3L
KLH conjugated synthetic peptide between 195-224 amino acids from the N-terminal region of human DI3L1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit