Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIAPH1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521409

Catalog No. PIPA521409


Add to cart

Description

Description

PA5-21409 targets Dia 1 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-21409 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 237 (O60610) of DIAPH1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

DIAPH1
Polyclonal
Unconjugated
DIAPH1
Liquid
DIAPH1
Recombit fragment corresponding to a region within amino acids 1 and 237 of DIAPH1 (Uniprot IDNo.O60610)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.69 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O60610
DFNA1, DIA1, DRF1, LFHL1, hDIA1, diaphanous-related formin 1, diaphanous-related formin-1, protein diaphanous homolog 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1729
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit