Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIABLO Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141509

Catalog No. 01-675-334


Add to cart

Description

Description

Suggested positive control: HeLa cell lysate.

Recently, three different laboratories have identified a molecule, Smac/DIABLO, which binds with inhibitor of apoptosis proteins (IAPs) and neutralizes IP inhibition of effector caspases. Multiple IAP molecules, including c-IAP1, c-IAP2, XIAP, and survivin are able to bind Smac. Smac/DIABLO is a mitochondrial protein, which is released into cytosol during apoptosis. The function of Smac is similar to that of Drosophila apoptosis activating proteins Reaper, Hid and Grim, which have been proposed to function by neutralizing the activity of IAPs. Smac promotes caspase-3 and caspase-9 activation by removing inhibitors rather than directly acting on Apaf-1, procaspases, and cytochrome c. The full-length cDNA of Smac encodes an open reading frame of 239 amino acids, predicting a protein with a molecular weight of 27 kDa. However, the mature protein purified from HeLa cells is only about 25 kDa (3).
Specifications

Specifications

DIABLO
Polyclonal
Unconjugated
DIABLO
Liquid
DIABLO
Synthetic peptide corresponding to residues 226-239 of human Smac.
Protein G
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
Q9NR28
hCG_1782202, DIABLO-S, FLJ10537, FLJ25049, SMAC, SMAC3
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
56616
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit