Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIABLO Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14267

Catalog No. PA14267


Add to cart

Description

Description

PA1-4267 detects DIABLO protein from human samples. PA1-4267 has been successfully used in Western blot procedures. PA1-4267 immunizing peptide corresponds to the C-terminus of the human DIABLO protein.

DIABLO (direct IAP binding protein with low pI, smac) is a novel protein that can bind mammalian IAP homolog A (MIHA, also known as XIAP), and can also interact with MIHB and MIHC and the baculoviral IAP, OpIAP. DIABLO may promote apoptosis by binding to IAPs and preventing them from inhibiting caspases.
Specifications

Specifications

DIABLO
Polyclonal
Unconjugated
DIABLO
Liquid
DIABLO
C-terminus of the human DIABLO protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9NR28
Smac, Second mitochondria-derived activator of caspase, Smac protein, Direct IAP-binding protein with low pI
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
56616
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit