Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIAPH1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527607

Catalog No. PIPA527607


Add to cart

Description

Description

PA5-27607 targets DIAPH1 in IF, IHC-P, and WB applications and shows reactivity with Human samples. The PA5-27607 immunogen is recombit fragment corresponding to a region within amino acids 981 and 1272 of Human DIAPH1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene is a homolog of the Drosophila diaphanous gene, and has been linked to autosomal domit, fully penetrant, nonsyndromic sensorineural progressive low-frequency hearing loss. Actin polymerization involves proteins known to interact with diaphanous protein in Drosophila and mouse. It has therefore been speculated that this gene may have a role in the regulation of actin polymerization in hair cells of the inner ear. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications

Specifications

DIAPH1
Polyclonal
Unconjugated
DIAPH1
Liquid
DIAPH1
Recombit fragment corresponding to a region within amino acids 981 and 1272 of Human DIAPH1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
O60610, O08808
DFNA1, DIA1, DRF1, LFHL1, hDIA1, diaphanous-related formin 1, diaphanous-related formin-1, protein diaphanous homolog 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
13367, 1729
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit