Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicer, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129791

Catalog No. PIPA129791

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Dicer is an RNase III-like enzyme which cleaves double-stranded RNA into 21-23 nt RNAs. These RNAs are referred to as siRNA (short interfering RNA). It is believed to be essential in siRNA-mediated gene suppression.

PA1-29791 detects Dicer from mouse samples.PA1-29791 has been successfully used in Western blot applications.The PA1-29791 immunogen is: Synthetic peptide (Mouse), corresponding to amino acids FTKHQVLIMTCYVALTVLK (132-150) and QNKQILVPELCAIHPI (1017 -1033).
Specifications

Specifications

Dicer
Polyclonal
whole serum diluted in PBS with 0.2% gelatin and 0.05% sodium azide
Q9UPY3, Q8R418
DCR1, Dicer, HERNA, KIAA0928
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Murine
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DICER1
Liquid
DICER1
Synthetic peptide (Mouse), corresponding to amino acids FTKHQVLIMTCYVALTVLK (132-150) and QNKQILVPELCAIHPI (1017 -1033).
100μg
Store at -20°C
Primary
192119, 23405
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit