Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dicer 1, ribonuclease type III, Mouse, Clone: 2F12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DICER1.
Supplier: Abnova Corporation H00023405M01
Description
This gene encodes a protein possessing an RNA helicase motif containing a DEXH box in its amino terminus and an RNA motif in the carboxy terminus. The encoded protein functions as a ribonuclease and is required by the RNA interference and small temporal RNA (stRNA) pathways to produce the active small RNA component that represses gene expression. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq
Sequence: ESLAGAIYMDSGMSLETVWQVYYPMMRPLIEKFSANVPRSPVRELLEMEPETAKFSPAERTYDGKVRVTVEVVGKGKFKGVGRSYRIAKSAAARRALRSLSpecifications
dicer 1, ribonuclease type III | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_177438 | |
DICER1 | |
DICER1 (NP_803187, 1813 a.a. ∼ 1912 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
50 μg | |
Angiogenesis | |
23405 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Western Blot | |
2F12 | |
Mouse monoclonal antibody raised against a partial recombinant DICER1. | |
DICER1 | |
DCR1/Dicer/HERNA/KIAA0928 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction