Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DIDO1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145947

Catalog No. NBP145947

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

DIDO1
Unconjugated
PBS, 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Affinity Purified
DIDO1
Synthetic peptide near the N-terminus of mouse Dio-1.
Protein A purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Western Blot
Western Blot
Q8C9B9
BYE1, C20orf158, DATF-1, DATF1DIDO3, death associated transcription factor 1, death inducer-obliterator 1, Death-associated transcription factor 1, death-inducer obliterator 1, DIO1DIDO2, DIO-1hDido1, dJ885L7.8, DKFZp434P1115, FLJ11265, KIAA0333chromosome 20 open reading frame 158, MGC16140
Rabbit
IgG
0.1 mg
Apoptosis
Primary
11083
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only