Learn More
Diethylstilbestrol Sheep anti-Chemical, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA175116
Catalog No. PIPA175116
Add to cart
Description
Description
PA1-75116 specifically detects Diethylstilbestrol. PA1-75116 has been successfully used in ELISA applications. The PA1-75116 immunogen is Diethylstilbestrol(p-OH)-BTG.
Diethylstilbestrol is a synthetic, nonsteroidal form of estrogen. A well-known teratogen and carcinogen, diethylstilbestrol inhibits the hypothalamic-pituitary-gonadal axis, thereby blocking the testicular synthesis of testosterone, lowering plasma testosterone, and inducing a chemical castration. Diethylstilbestrol is used in the treatment of menopausal and postmenopausal disorders. It was also used formerly as a growth promoter in animals. [National Center for Biotechnology Information. PubChem Compound Database; CID=448537]
Specifications
Specifications
Diethylstilbestrol
Polyclonal
PBS with 0.09% sodium azide
Diethylstilbestrol; Stilbestrol; Stilboestrol; Distilbene; C18H20O2
Diethylstilbestrol(p-OH)-BTG
Purified
RUO
Antibody
Polyclonal
ELISA
Unconjugated
Liquid
Sheep
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical