Learn More
DHDH Mouse anti-Human, Mammalia, Mouse, Porcine, Clone: T101, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112599
Catalog No. 01-674-367
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody recognizes the DD1 or DD2 isoforms. In Western blot, this antibody detects a ∽32 kDa band in human breast cancer tissue and porcine liver tissue.
DDH is a member of aldo-keto reductase superfamily, which catalyzes reduction of aldehyde or ketone to a corresponding alcohol by using NADH or NADPH as a cofactor. In liver, the enzyme is abundantly located in the cytoplasm as a monomeric 34–36 kDa protein.
Specifications
Specifications
DHDH
Monoclonal
Unconjugated
DHDH
Liquid
DHDH, zgc:101723
Recombit type 2 dihydrodiol dehydrogenase from human lung cancer
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100008610, 27294, 397337, 403786, 444539, 449558, 617564, 691002, 71755
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
T101
ascites diluted in PBS with 0.02% sodium azide
Q148L6, Q9TV68, Q9UQ10, Q9DBB8, Q9TV69, Q9TV70, Q6DKE0, Q642M9
DDH
Mouse
IgG1
RUO
Antibody
Monoclonal
Human, Mammalia, Mouse, Porcine