Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl-Histone H3 (Arg2) Rabbit anti-Human, Vertebrates, Clone: 20.2, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04808
Description
Use Anti-dimethyl-Histone H3 (Arg2), clone 20.2 Antibody (rabbit monoclonal antibody) validated in DB, Mplex, PIA, western blotting to detect dimethyl-Histone H3 (Arg2) also known as H3R2me2, Histone H3 (di methyl R2).Specifications
dimethyl-Histone H3 (Arg2) | |
Monoclonal | |
Unconjugated | |
NM_003493 | |
KLH-conjugated, synthetic peptide containing the sequence ...Ame2RTKQ..., in which me2R corresponds to dimethyl-arginine 2 of human histone H3. | |
Primary | |
Human, Vertebrates | |
Supernatant |
Multiplex, Immunoassay, Inhibition Assays, Western Blot | |
20.2 | |
Cultured supernantant in 0.05% sodium azide | |
Rabbit | |
200 μL | |
HIST3H3, H3FT, MGC126886, H3t, MGC126888, H3T, H3/g, H3.4, H3/t | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only