Learn More
Dimethyl-Histone H3 (Lys9) Rabbit anti-Human, Clone: 55-10C-1, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05768R
Catalog No. 05-768-RMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-dimethyl-Histone H3 (Lys9) Antibody (Rabbit Monoclonal Antibody) validated in western blot, PIA to detect dimethyl-Histone H3 (Lys9) also known as H3K9me2, Histone H3 (di methyl K9).
Specifications
Specifications
dimethyl-Histone H3 (Lys9)
55-10C-1
Purified Rabbit Monoclonal IgG Supernatant in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) containing 40% glycerol with 0.05% sodium azide.
Purified
KLH- Conjugated - Linear Peptide corresponding to the N-terminus of Dimethyl-Histone H3 (Lys9). Methylated at and around Lysine 9
Protein A purfied
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Monoclonal
Human
Inhibition Assays, Western Blot
Unconjugated
NM_003493.2
Rabbit
IgG
100μL
Primary
HIST3H3, H3FT, MGC126886, H3t, MGC126888, H3T, H3/g, H3.4, H3/t