Learn More
anti-Dimethyl Histone H3 (Lys9), Clone: CMA307, EMD Millipore™ Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Millipore Sigma 051249
 View more versions of this product
Catalog No. 05-124-9MI
  Check Availability
Add to cart
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
Dimethyl Histone H3 (Lys9)
CMA307
Mouse monoclonal IgG1κ supernatant in PBS with 0.05% sodium azide.
Purified
Murine
κ
100μg
Primary
Human,Vertebrate
ELISA,Immunocytochemistry,Immunoprecipitation,Western Blot
Unlabeled
NM_003493
H3.4, H3/g , H3/t , H3FT , H3T, H3t , MGC126886, MGC126888 , OTTHUMP00000037945
Synthetic peptide corresponding to amino acids 1-19 of human Histone H3, dimethylated on Lys9, conjugated to KLH.
Protein G purfied
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal