Learn More
anti-dimethyl-Histone H3 (Lys9), Polyclonal, Millipore™ (Upstate™)
Manufacturer:  Millipore Sigma 07441/DEL
 View more versions of this product
Catalog No. 50-172-446
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
dimethyl-Histone H3 (Lys9)
Semi-purified
KLH-conjugated, 2X-branched synthetic peptide containing the sequence ..Rme2KSTG.., in which me2K corresponds to dimethyl-lysine at residue 9 of human Histone H3
RUO
2 years at -20°C
Polyclonal
Unlabeled
Rabbit
IgG
Epigenetics
Primary
Chicken,Human,Murine,Rat,Yeast