Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl-Histone H4 (Arg3) Asymmetric (Trial Size) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$135.00 - $553.00
Specifications
Antigen | dimethyl-Histone H4 (Arg3) Asymmetric |
---|---|
Applications | Dot Blot, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
07-213-ISMI
|
MilliporeSigma
07213IS |
10 μL |
Each for $135.00
|
|
|||||
07-213-IMI
|
MilliporeSigma
07213I |
100 μL |
Each for $553.00
|
|
|||||
Description
This Anti-dimethyl-Histone H4 (Arg3) Asymmetric Antibody is validated for use in Western Blotting and Immunoprecipitation and Dot Blot for the detection of dimethyl-Histone H4 (Arg3) Asymmetrimmunocytochemistry.Specifications
dimethyl-Histone H4 (Arg3) Asymmetric | |
Polyclonal | |
Rabbit | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HIST1H4A; H4/A; H4FA; HIST1H4B; H4/I; H4FI; HIST1H4C; H4/G; H4FG; HIST1H4D; H4/B; H4FB; HIST1H4E; H4/J; H4FJ; HIST1H4F; H4/C; H4FC; HIST1H4H; H4/H; H4FH; HIST1H4I; H4/M; H4FM; HIST1H4J; H4/E; H4FE; HIST1H4K; H4/D; H4FD; HIST1H4L; H4/K; H4FK; HIST2H4A; H4/N; H4F2; H4FN; HIST2H4; HIST2H4B; H4/O; | |
Primary | |
2-8°C for 12 months |
Dot Blot, Immunoprecipitation, Western Blot | |
Unconjugated | |
Human, Mouse, Rat | |
NP_001029249 | |
KLH-conjugated linear peptide corresponding to the N-terminus of human dimethyl-Histone H4 (Arg3) Asymmetric. | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title