Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIO3 Rabbit anti-Hamster, Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522962

Catalog No. PIPA522962


Add to cart

Description

Description

The target sequence has 93% sequence homology with human. Suggested positive control: rat placenta tissue lysate.

Dio3, or type 3 iodothyronine deiodinase, can regulate circulating fetal thyroid hormone concentrations throughout gestation. It has an essential role for regulation of thyroid hormone inactivation during embryological development, and may play a role in preventing premature exposure of developing fetal tissues to adult levels of thyroid hormones. Dio3 has been linked to the development of consumptive hypothyroidism in both infants and adults.
Specifications

Specifications

DIO3
Polyclonal
Unconjugated
DIO3
Liquid
DIO3
Synthetic peptide made to an internal portion of the rat protein (within residues 250-300).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse, Rabbit, Rat
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 30% glycerol and 0.1% sodium azide
P55073, Q91ZI8, P49897
5DIII, DIOIII, type 3 DI, type III iodothyronine deiodinase, type-III 5′-deiodinase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100357267, 100753739, 107585, 1735, 29475
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit