Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIRAS1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526409

Catalog No. PIPA526409

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DIRAS1 belongs to a distinct branch of the functionally diverse Ras (see HRAS; MIM 190020) superfamily of monomeric GTPases.
Specifications

Specifications

DIRAS1
Polyclonal
PBS with 0.09% sodium azide
O95057
Di-Ras1, GBTS1, RIG, GTP-binding protein Di-Ras1, distinct subgroup of the Ras family member 1, ras-related inhibitor of cell growth, small GTP-binding tumor suppressor 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
148252
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DIRAS1
Liquid
DIRAS1
KLH conjugated synthetic peptide between 109-136 amino acids from the central region of human DIRAS1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit