Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heartworm Goat anti-Nematode, HRP, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA173111

Catalog No. PIPA173111


Add to cart

Description

Description

The PA1-73111 antibody reacts with Dirofiliaria immitis. PA1-73111 has been successfully used in ELISA applications. The PA1-73111 antibody was raised against Dirofiliaria immitis adult worm preparation.

Canine heartworm is a common parasite of dogs worldwide. Heartworm occurs in about 30 other species as well but rarely affects humans. The infection is commonly transmitted by mosquitoes. Adult heartworms can grow to be up to 10 inches in length and can live for 5-7 years in dogs affecting the heart and lungs.
Specifications

Specifications

Canine Heartworm
Polyclonal
HRP
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Nematode
ELISA
1-2 mg/mL
PBS with 10mg/mL BSA and 0.002% thimerosal
Adult canine heartworm; Filaria; dirofilariasis
Dirofiliaria immitis adult worm preparation.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit