Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIS3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523705

Catalog No. PIPA523705


Add to cart

Specifications

Specifications

DIS3
Polyclonal
PBS with 0.09% sodium azide
Q9Y2L1, Q9CSH3
EXOSC11, KIAA1008, RP11-342J4.3, RRP44, bA555G22.1, dis3p, 2810028N01Rik, exosome complex exonuclease RRP44, exosome component 11, mitotic control protein dis3 homolog, protein DIS3 homolog, ribosomal RNA-processing protein 44
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22894, 72662
Immunohistochemistry, Western Blot
Unconjugated
DIS3
Liquid
DIS3
KLH conjugated synthetic peptide between 295-323 amino acids from the central region of human DIS3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit