Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DISC1 Polyclonal Antibody, HRP, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522839
Description
The theoretical molecular weight of DISC1 is ∽90 kDa. Due to alternative splicing, DISC1 may also run at ∽75 and 100 kDa. Suggested positive control: human brain sections (neurons and glia).
DISC1 Polyclonal antibody specifically detects DISC1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
DISC1 | |
Polyclonal | |
HRP | |
DISC1 | |
C1orf136; DISC1; disrupted in schizophrenia 1; disrupted in schizophrenia 1 homolog; Disrupted in schizophrenia 1 protein; KIAA0457; RP4-730B13.1; SCZD9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
244667, 27185, 307940 | |
4° C, store in dark | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
0.85 mg/mL | |
PBS with no preservative | |
Q810H6, Q811T9, Q9NRI5 | |
DISC1 | |
A synthetic peptide made to an internal region (within residues 400-500) of the rat DISC1 protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction