Learn More
Disialoganglioside GD2 Mouse anti-Human, PerCP-Cy5.5, Clone: 14.G2a, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563438
Catalog No. BDB563438
Add to cart
Description
Description
Gangliosides are sialic-acid bearing glycolipids that are expressed on the surface of all mammalian cells, and are likely involved in mediating cell-substratum interactions. They are important target antigens for antibody dependent cellular cytotoxicity (ADCC) of human melanoma and neuroblastoma cells. Human melanoma cells produce gangliosides, designated as GD2 and GD3 which are deposited in the subtratum-attached material, and may play a significant role in the melanoma metastatic phenotype. Clone 14.G2a specifically reacts with human and mouse GD2 ganglioside. LAN-1 human neuroblastoma cells were used as immunogen. Clone 14.G2a is an isotype switch variant selected from the parental IgG3-producing hybridoma 14.18 and has identical reactivity as the parental antibody. Clone 14.G2a is routinely tested by flow cytometry using M21 human melanoma cells.
Specifications
Specifications
Disialoganglioside GD2
14.G2a
GD2
Affinity Purified
LAN-1 human neuroblastoma cells
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PerCP-Cyanine5.5
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Primary
Human