Learn More
Disopyramide Mouse anti-Chemical, Clone: DS13-788.1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110721
Catalog No. 01-671-951
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10721 detects Disopyramide. MA1-10721 has been successfully used in ELISA applications. The MA1-10721 immunogen is Disopyramide, conjugated to KLH.
Disopyramide is a class I anti-arrhythmic agent (one that interferes directly with the depolarization of the cardiac membrane and thus serves as a membrane-stabilizing agent) with a depressant action on the heart similar to that of guanidine. It also possesses some anticholinergic and local anesthetic properties. [National Center for Biotechnology Information. PubChem Compound Database; CID=3114]
Specifications
Specifications
Disopyramide
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Norpace; Rythmodan; Disopyramide; Dicorantil; Ritmodan
Disopyramide, conjugated to KLH.
Purified
RUO
Antibody
Monoclonal
ELISA
DS13-788.1
Unconjugated
Liquid
Mouse
IgG2b
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical