Learn More
DJ-1 Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA146262
Catalog No. PA146262
Add to cart
Description
Description
This antibody is predicted to react with rat, bovine, zebra fish and Golden Syrian hamster based on 100% sequence homology. Suggested positive control: antigen standard for PARK7 (transient overexpression lysate).
DJ-1 is one of several genes thought to cause autosomal-recessive early-onset parkinsonism.
Specifications
Specifications
DJ-1
Polyclonal
whole serum with no preservative
Q8UW59, Q99497, Q99LX0, O88767
CTA-215D11.1, DJ-1, DJ1, FLJ27376, FLJ34360, FLJ92274
Rabbit
IgG
RUO
Antibody
Polyclonal
Chicken, Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In vitro Assay, Western Blot
Unconjugated
PARK7
Liquid
PARK7
Residues between 150-189 of human DJ-1 (the C-terminus).
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11315, 117287, 395277, 57320