Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDHR1, Polyclonal, Invitrogen™

Catalog No. PIPA127522
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127522 50 μg
1 options

Catalog No. PIPA127522

Supplier: Thermo Scientific PA127522

Chicken Polyclonal Antibody

This protein is a member of the cadherin superfamily of calcium-dependent cell-cell adhesion molecules. The encoded protein has a signal peptide. six cadherin repeat domains and a unique cytoplasmic region. This non-classical cadherin appears to be exclus

PA1-27522 detects DKFZp434A132 from human samples.PA1-27522 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27891 immunogen is a synthetic peptide corresponding to human Hypothetical protein DKFZp434A132.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DKFZp434A132
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene CDHR1
Gene Accession No. Q96JP9
Gene Alias DKFZp434A132, KIAA1775, PCDH21, PRCAD
Gene Symbols CDHR1
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Hypothetical protein DKFZp434A132.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92211
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.