Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

immunoglobulin-like and fibronectin type III domain containing 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant DKFZp434B1231.

Manufacturer:  Abnova Corporation H00091156A01

Catalog No. 89-004-759


Add to cart

Description

Description

Sequence: GILPGHEYHFRVVAKNELGASKPSDTSQPWCIPRQRDRFTVKAPCYREPDLSQKPRFLVGLRSHLLPQGCECCMSCAVQGSPRPHVTWFKNDRSLEGNP
Specifications

Specifications

immunoglobulin-like and fibronectin type III domain containing 1
Unconjugated
50% glycerol
NM_178275
DKFZp434B1231|EEF1A2BP1
Mouse
50 uL
Cell Adhesion
Y
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant DKFZp434B1231.
IGFN1
Serum
IGFN1
DKFZp434B1231 (NP_840059, 720 a.a. ~ 818 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
91156
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit