Learn More
Aviva Systems Biology Corp ANTI-DKFZP779O175 50UG/50ULAN DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP50425_P050
This is a rabbit polyclonal antibody against DKFZp779O175. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DKFZp779O175. Alternate names: DKFZp686K21248, FLJ27459, MGC129866, MGC129867, DKFZp779O175ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-53841