Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Dkk-2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145520

Catalog No. NBP145520

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Dkk-2
0.5 mg/mL
Western Blot
NP_064661.2
dickkopf (Xenopus laevis) homolog 2, dickkopf 2, dickkopf homolog 2 (Xenopus laevis), dickkopf related protein-2, dickkopf-2, dickkopf-related protein 2, DKK-2, hDkk-2
Rabbit
IgG
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Western Blot
Unconjugated
PBS, 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Affinity Purified
DKK2
Synthetic peptide surrounding amino acid 241 of mouse Dkk2 (.
Immunogen affinity purified
RUO
Primary
27123
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only