Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DKK3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521325

Catalog No. PIPA521325


Add to cart

Description

Description

PA5-21325 targets Dkk-3 in ICC, IF, and WB applications and shows reactivity with Human and mouse samples. The PA5-21325 immunogen is recombit protein fragment corresponding to a region within amino acids 6 and 343 (Q9UBP4) of DKK3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

DKK3
Polyclonal
Unconjugated
DKK3
Liquid
DKK3
Recombit protein fragment corresponding to a region within amino acids 6 and 343 (Q9UBP4) of DKK3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q9UBP4, Q9QUN9
UNQ258/PRO295, REIC, RIG, RIG-like 5-6, RIG-like 7-1, dickkopf homolog 3, dickkopf-3, dickkopf-related protein 3, dkk-3, hDkk-3, regulated in glioma
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
27122, 50781
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit