Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DKK1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523187

Catalog No. PIPA523187


Add to cart

Description

Description

DKK1 is a 266 amino acid secreted protein with two cysteine rich regions and belonging to the dickkopf family. It is involved in embryonic development through its inhibition of the WNT signaling pathway and in the reentry into the cell cycle of human adult stem cells from bone marrow. Its loss of expression plays a role in development or progression of maligt melanoma. It decreases melanocyte function and is a key player in multiple myeloma bone disease. It is a potential prognostic and diagnostic marker for breast cancer patients with poor prognosis.
Specifications

Specifications

DKK1
Polyclonal
Unconjugated
DKK1
Liquid
DKK1
Synthetic peptide (167 GYSRRTTLSSKMYHTKGQEGSVC 185) of DKK1 protein.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
O94907, O54908
UNQ492/PRO1008, DKK-1, SK, dickkopf related protein-1, dickkopf-1 like, dickkopf-related protein 1, hDkk-1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
13380, 22943
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit