Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dlg Mouse, Unlabeled, Clone: 12, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610875

 View more versions of this product

Catalog No. BDB610875


Add to cart

Description

Description

The human homologue of the Drosophila discs large tumor suppressor protein (hDlg) is a member of the MAGUKs (membrane associated guanylate kinases) protein family. Members of this family (PSD-95, ZO-1, ZO-2, and human erythroid p55) are involved in cell structure and signaling events. The hDlg protein consists of several domains: three PDZ (PSD-95/Discs large/ZO-1) domains, an SH3 domain, and a guanylate kinase-like domain. However, hDlg contains a proline rich N-terminus region consisting of two SH3 domain binding sites that are not normally found in the MAGUKs family. The PDZ domains mediate the interaction of several proteins, such as Shaker-type K+ channel proteins and the APC tumor suppressor protein. Dlg is a peripheral membrane that associates with the cytoskeleton. The cellular location and binding sites of Dlg suggest a role in structure, signal transduction, and growth regulation. Supporting these probable Dlg functions are reports demonstrating that recessive mutations in Drosophila dlg lead to imaginal disc neoplasia and death. Also, Dlg has been reported to bind p56 [lck] tyrosine kinase and the Kv1.3 channel in human T lymphocytes.
Specifications

Specifications

Dlg
12
Unconjugated
Affinity Purified
Human Dlg aa. 5-213
150μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.