Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-DLG1, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP120907
Description
DLG1 Polyclonal specifically detects DLG1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
DLG1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_001091894.1and NP_004078.2). | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_001091894.1 | |
DLG1 | |
Peptide with sequence C-NSKTRDKGEIPDD corresponding to internal region according to NP_001091894.1, NP_004078.2. | |
0.1 mg | |
Vision | |
1739 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only