Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLG7, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127490

Catalog No. PA127490

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Potential cell cycle regulator that may play a role in carcinogenesis of cancer cells. Mitotic phosphoprotein regulated by the ubiquitin-proteasome pathway. Key regulator of adherens junction integrity and differentiation that may be involved in CDH1-medi

PA1-27490 detects DLG7 from human samples.PA1-27490 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27556 immunogen is a synthetic peptide corresponding to human Discs large homolog 7.Store at 4°C short term; for long term storage, store at -20°C, avoiding freeze/thaw cycles.
Specifications

Specifications

DLG7
Polyclonal
PBS with no preservative
Q15398
DLG7, HURP, KIAA0008
Chicken
IgY
50μg
Store at -20°C
Primary
9787
Western Blot
Unconjugated
DLGAP5
Purified
DLGAP5
Synthetic peptide corresponding to human Discs large homolog 7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit