Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLL1 Mouse anti-Human, Clone: 251127, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen DLL1
Immunogen Mouse myeloma cell line NS0-derived recombinant human DLL1 Ser22-Gly540 Accession # AAG09716
Clone 251127
Applications Western Blot, Flow Cytometry, CyTOF, ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1818SP
View Documents
R&D Systems
MAB1818SP
25 ug
Each for $114.50
Add to cart
 
MAB1818100
View Documents
R&D Systems
MAB1818100
100 ug
Each for $377.00
Add to cart
 
MAB1818500
View Documents
R&D Systems
MAB1818500
500 ug
Each for $839.00
Add to cart
 
Description

Description

DLL1 Monoclonal antibody MAB1818 Specifically detects DLL1 in Human samples. It is validated for ELISA Capture, Flow Cytometry, Western Blotting
Specifications

Specifications

DLL1
251127
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2b
28514
RUO
Mouse myeloma cell line NS0-derived recombinant human DLL1 Ser22-Gly540 Accession # AAG09716
Western Blot, Flow Cytometry, CyTOF, ELISA
Purified
AAG09716
Monoclonal
Primary
Detects recombinant human DLL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse DLL4 is observed.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit