Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-DLL1 Clone: 4F8,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ ST1703100UG
Description
Specifically detects DLL1 Clone: 4F8 in Human samples, and it is validated for ELISA, Immunocytochemistry, Immuno Blotting
The DLL1 protein is a human homolog of the Notch Δ ligand and is a member of the δ/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis. It may also play a role in cell-to-cell communication.Specifications
DLL1 | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human | |
Purified |
ELISA, Immunoblot, Immunoblot, Immunocytochemistry | |
4F8 | |
In PBS, pH 7.2. | |
A recombinant polypeptide corresponding to amino acids of 18-110 human DLL1, expressed as a GST fusion protein | |
RUO | |
Recognizes the DLL1 protein in HeLa cells. | |
−20°C | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction