Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DLL1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP126400

Catalog No. NBP126400

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

DLL1
0.5 mg/mL
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000
O00548
Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1
Goat
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Rat. Expected from sequence similarity: Human, Mouse .
Western Blot, ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
DLL1
Peptide with sequence C-ATQRHLTVGEEWSQD corresponding to internal region according to NP_005609.3.
78 kDa
0.1 mg
Neuroscience, Stem Cell Signaling Pathway
Primary
28514
Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only