Learn More
anti-DLL4, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP131195
Catalog No. NBP131195
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
DLL4
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
DLL4
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human DLL4. The exact sequence is proprietary.
75 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
54567
Human