Learn More
DLX1 Mouse anti-Human, Clone: 1D11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518735
Catalog No. PIMA518735
Add to cart
Description
Description
Peptide Sequence: YLALPERAEL AASLGLTQTQ VKIWFQNKRS KFKKLMKQGG AALEGSALAN GRALSAGSPP VPPGWNPNSS SGKGSGGNAG SYIPSYTSWY PSAHQEAMQQ PQLM
This gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. The encoded protein is localized to the nucleus where it may function as a transcriptional regulator of signals from multiple TGF-{beta} superfamily members. The encoded protein may play a role in the control of craniofacial patterning and the differentiation and survival of inhibitory neurons in the forebrain. This gene is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 2. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
Specifications
Dlx1
Monoclonal
0.43 mg/mL
PBS with no preservative; pH 7.4
P56177
distal-less homeo box 1, homeobox protein DLX-1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1745
ELISA, Immunoprecipitation, Western Blot
1D11
Unconjugated
DLX1
Liquid
DLX1
DLX1 (NP_835221, 152 a.a. ∽ 255 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human