Learn More
DLX2 Mouse anti-Human, Clone: 3H3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518739
Catalog No. PIMA518739
Add to cart
Description
Description
Peptide Sequence: MTGVFDSLVA DMHSTQIAAS STYHQHQQPP SGGGAGPGGN SSSSSSLHKP QESPTLPVST ATDSSYYTNQ QHPAGGGGGG GSPYAHMGSY QYQASGLNNV PYSAKSSYDL GYTAAYTSYA PYGT
Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. This gene is located in a tail-to-tail configuration with another member of the gene family on the long arm of chromosome 2.
Specifications
Specifications
DLX2
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q07687
Distal-less homeobox 2, TES-1, TES1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1746
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
3H3
Unconjugated
DLX2
Liquid
DLX2
DLX2 (NP_004396, 1 a.a. ∽ 124 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human